Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSTR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001053.3 | NP_001044.1 | 1 | 312 | ||
NM_001172560.1 | NP_001166031.1 | 2 | 295 | |||
GenBank mRNA | AK308381.1 | - | 1 | 312 | ||
DB202230.1 | - | 1 | 312 | |||
DQ448303.1 | - | 1 | 224 | |||
DQ448304.1 | - | 1 | 224 |
Set Membership: |
G-protein coupled receptor |