Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLA2G1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000928.2 | NP_000919.1 | 3 | 248 | ||
GenBank mRNA | AK311830.1 | - | 3 | 233 | ||
BC106725.1 | - | 3 | 248 | 40014551 | ||
BC106726.1 | - | 3 | 248 | 40014552 | ||
BE969737.1 | - | 219 | ||||
BX113838.1 | - | 2 | 52 | |||
CA867923.1 | - | 2 | 91 | |||
M21054.1 | - | 3 | 249 |