Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGT2B17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077.3 | NP_001068.1 | 1 | 480 | ||
GenBank mRNA | U59209.1 | - | 1 | 488 |