Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSC22D3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015881.1 | NP_001015881.1 | 3 | 297 | ||
NM_001318468.1 | NP_001305397.1 | 4 | 765 | |||
NM_001318470.1 | NP_001305399.1 | 4 | 751 | |||
NM_004089.3 | NP_004080.2 | 3 | 502 | |||
NM_198057.2 | NP_932174.1 | 3 | 818 | |||
XM_005262099.1 | XP_005262156.1 | 5 | 779 | |||
XM_005262100.1 | XP_005262157.1 | 4 | 654 | |||
XM_005262102.1 | XP_005262159.1 | 4 | 647 | |||
XM_005262103.3 | XP_005262160.1 | 4 | 757 | |||
XM_011530884.1 | XP_011529186.1 | 4 | 853 | |||
XM_017029335.1 | XP_016884824.1 | 4 | 589 | |||
GenBank mRNA | AB025432.1 | - | 3 | 485 | ||
AF153603.1 | - | 3 | 480 | |||
AF183393.1 | - | 3 | 488 | |||
AF228339.1 | - | 3 | 492 | |||
AK092645.1 | - | 4 | 704 | |||
AK092669.1 | - | 3 | 790 | |||
AK127938.1 | - | 3 | 442 | |||
AK300535.1 | - | 4 | 629 | |||
AK311834.1 | - | 3 | 397 | |||
AL110191.1 | - | 3 | 501 | |||
AY007119.1 | - | 3 | 401 | |||
BC018148.1 | - | 3 | 184 | |||
BC072446.1 | - | 3 | 496 | 6165765 | ||
BQ919216.1 | - | 297 | ||||
BX647854.1 | - | 4 | 751 | |||
CR533450.1 | - | 3 | 240 | |||
CR933650.1 | - | 3 | 504 | |||
Z50781.1 | - | 3 | 216 |