Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTTG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282382.1 | NP_001269311.1 | 5 | 1037 | ||
NM_001282383.1 | NP_001269312.1 | 6 | 891 | |||
NM_004219.3 | NP_004210.1 | 6 | 730 | |||
GenBank mRNA | AF062649.1 | - | 6 | 701 | ||
AF095287.1 | - | 6 | 670 | |||
AJ223953.1 | - | 5 | 739 |