Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FXYD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022006.1 | NP_071289.1 | 3, 4 | 229 | ||
GenBank mRNA | AK057825.1 | - | 3, 4 | 204 | ||
BC018619.1 | - | 3, 4 | 213 | 3626060 |