Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033504.2 | NP_277039.1 | 2 | 290 | ||
XM_005270408.3 | XP_005270465.1 | 2 | 373 | |||
GenBank mRNA | AY027543.1 | - | 2 | 227 | ||
AY359883.1 | - | 2 | 176 | |||
BC001418.3 | - | 2 | 290 | 3140820 | ||
BC013953.2 | - | 2 | 290 | 2987806 | ||
BC023260.1 | - | 2 | 327 | 4213210 |