Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318015.1 | NP_001304944.1 | 5 | 1273 | ||
NM_001318016.1 | NP_001304945.1 | 5 | 1136 | |||
NM_001318020.1 | NP_001304949.1 | 5 | 1086 | |||
NM_001318023.1 | NP_001304952.1 | 5 | 1105 | |||
NM_032799.4 | NP_116188.2 | 5 | 1108 | |||
XM_011519116.2 | XP_011517418.1 | 5 | 1260 | |||
GenBank mRNA | AK027430.1 | - | 6 | 945 | ||
AK075332.1 | - | 6 | 945 | |||
AK092843.1 | - | 4 | 2503 | |||
BC009280.2 | - | 5 | 1085 | 4302257 | ||
BC048251.1 | - | 5 | 1254 | 5418875 |