Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MYB SIRNA Assays ›
Gene Symbol:
MYB
Gene Name:
MYB proto-oncogene, transcription factor
Gene Aliases:
Cmyb, c-myb, c-myb_CDS, efg
Chromosome Location:
Chr.6: 135180981 - 135219173 on Build GRCh38
UniGene ID:
Hs.606320
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
42848
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MYB
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130172.1
NP_001123644.1
5
576
NM_001130173.1
NP_001123645.1
5
576
NM_001161656.1
NP_001155128.1
5
576
NM_001161657.1
NP_001155129.1
5
576
NM_001161658.1
NP_001155130.1
5
576
NM_001161659.1
NP_001155131.1
5
576
NM_001161660.1
NP_001155132.1
5
576
NM_005375.3
NP_005366.2
5
576
NR_134958.1
5
576
NR_134959.1
5
576
NR_134960.1
5
576
NR_134961.1
5
576
NR_134963.1
5
576
NR_134964.1
5
576
NR_134965.1
5
576
GenBank mRNA
AF104863.1
-
5
454
AJ606317.1
-
5
576
AJ606318.1
-
5
576
AJ606319.1
-
5
576
AJ606320.1
-
5
576
AJ606321.1
-
5
576
AJ606322.1
-
5
576
AJ606323.1
-
5
576
AJ606324.1
-
5
576
AJ616235.1
-
5
491
AJ616795.1
-
1
71
AY787443.1
-
5
377
AY787445.1
-
4
305
AY787447.1
-
5
377
AY787448.1
-
5
377
AY787449.1
-
5
377
AY787450.1
-
5
377
AY787451.1
-
5
377
AY787452.1
-
5
377
AY787453.1
-
5
377
AY787454.1
-
5
377
AY787455.1
-
5
377
AY787456.1
-
5
377
AY787457.1
-
5
377
AY787458.1
-
5
377
AY787459.1
-
5
377
AY787460.1
-
5
377
AY787461.1
-
5
377
AY787462.1
-
5
377
AY787463.1
-
5
377
AY787464.1
-
5
377
AY787465.1
-
5
377
AY787466.1
-
5
377
AY787467.1
-
5
377
AY787468.1
-
5
377
AY787469.1
-
5
377
AY787470.1
-
5
377
AY787471.1
-
5
377
AY787472.1
-
5
377
AY787473.1
-
5
377
AY787474.1
-
5
377
AY787475.1
-
5
377
BC064955.1
-
5
528
6069320
M13665.1
-
3
239
M13666.1
-
3
239
M15024.1
-
5
490
X52125.1
-
5
574
X52126.1
-
4
380
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
mitotic cell cycle
chromatin remodeling
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
positive regulation of T-helper cell differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of histone H3-K4 methylation
positive regulation of histone H3-K9 methylation
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, RNA polymerase II transcription factor recruiting
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-7dvbj:80/10.239.167.51:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0