Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002467.4 | NP_002458.2 | 2 | 719 | ||
GenBank mRNA | AK303921.1 | - | 2, 3 | 501 | ||
AK312883.1 | - | 2 | 552 | |||
BC000141.1 | - | 2 | 530 | 2985844 | ||
BC000917.2 | - | 2 | 546 | 3048750 | ||
BC058901.1 | - | 2 | 232 | 6012670 | ||
BT019768.1 | - | 1 | 149 | |||
K02276.1 | - | 2 | 569 | |||
M13929.1 | - | 3 | 662 | |||
M13930.1 | - | 2 | 1353 | |||
V00568.1 | - | 2 | 707 |