Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178026.2 | NP_821158.2 | 15 | 1932 | ||
XM_011528782.2 | XP_011527084.1 | 13 | 1411 | |||
XM_011528783.2 | XP_011527085.1 | 10 | 1180 | |||
XM_017027809.1 | XP_016883298.1 | 15 | 1924 | |||
GenBank mRNA | BC141843.1 | - | 15 | 1921 | 8860314 | |
DA117929.1 | - | 2 | 172 |