Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STRADA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003786.2 | NP_001003786.1 | 11 | 1843 | ||
NM_001003787.2 | NP_001003787.1 | 13 | 1954 | |||
NM_001003788.2 | NP_001003788.1 | 12 | 1925 | |||
NM_001165969.1 | NP_001159441.1 | 11 | 1776 | |||
NM_001165970.1 | NP_001159442.1 | 11 | 1731 | |||
NM_153335.5 | NP_699166.2 | 9 | 2389 | |||
XM_005257797.2 | XP_005257854.1 | 13 | 1805 | |||
XM_005257798.2 | XP_005257855.1 | 11 | 1767 | |||
XM_005257799.2 | XP_005257856.1 | 12 | 1776 | |||
XM_005257800.2 | XP_005257857.1 | 13 | 1738 | |||
XM_011525466.2 | XP_011523768.1 | 12 | 1786 | |||
XM_011525467.2 | XP_011523769.1 | 13 | 1856 | |||
XM_017025312.1 | XP_016880801.1 | 11 | 1627 | |||
XM_017025313.1 | XP_016880802.1 | 12 | 1709 | |||
GenBank mRNA | AF308302.1 | - | 13 | 1793 | ||
AK074771.1 | - | 11 | 1709 | |||
AK075005.1 | - | 9 | 2223 | |||
AK128173.1 | - | 1 | 3309 | |||
AL832407.1 | - | 12 | 1738 | |||
AY290821.2 | - | 9 | 2389 | |||
BC043641.1 | - | 10 | 2577 | 3908187 | ||
BQ682133.1 | - | 1 | 224 |