Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAQR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005262769.4 | XP_005262826.1 | 5, 6 | 990 | ||
XM_017007785.1 | XP_016863274.1 | 5, 6 | 990 | |||
GenBank mRNA | BC047510.1 | - | 5, 6 | 965 | 5297045 |