Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBR7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175748.3 | NP_786924.2 | 1, 2 | 394 | ||
GenBank mRNA | AK097477.1 | - | 1, 2 | 379 | ||
AK315492.1 | - | 1, 2 | 193 | |||
BC015046.1 | - | 1, 2 | 193 | 3908032 | ||
BC051819.1 | - | 1, 2 | 156 | 6171393 | ||
BX248249.1 | - | 1, 2 | 153 | |||
DB047433.1 | - | 1, 2 | 394 |