Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAS2R20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176889.2 | NP_795370.2 | 1 | 477 | ||
GenBank mRNA | BC100915.2 | - | 1 | 477 | 40003843 | |
BC100916.2 | - | 1 | 477 | 40003844 | ||
BC100917.2 | - | 1 | 477 | 40003847 | ||
BC100918.2 | - | 1 | 477 | 40003851 | ||
BC113843.1 | - | 1 | 477 | 40003846 | ||
BC114451.1 | - | 1 | 477 | 40003845 |