Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MT1E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005255956.4 | XP_005256013.1 | 2 | 425 | ||
GenBank mRNA | AF495759.1 | - | 2 | 311 |