Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ANKMY1 SIRNA Assays ›
Gene Symbol:
ANKMY1
Gene Name:
ankyrin repeat and MYND domain containing 1
Gene Aliases:
ZMYND13
Chromosome Location:
Chr.2: 240468632 - 240561110 on Build GRCh38
UniGene ID:
Hs.656615
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
44256
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ANKMY1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282771.1
NP_001269700.1
3, 4
502
NM_001282780.1
NP_001269709.1
3, 4
502
NM_001282781.1
NP_001269710.1
3, 4
502
NM_001308375.1
NP_001295304.1
3, 4
741
NM_016552.3
NP_057636.2
2, 3
339
NM_017844.3
NP_060314.2
3, 4
502
XM_011511300.2
XP_011509602.1
3, 4
686
XM_011511301.1
XP_011509603.1
3, 4
686
XM_011511304.2
XP_011509606.1
3, 4
686
XM_011511305.2
XP_011509607.1
3, 4
686
XM_011511309.2
XP_011509611.1
3, 4
686
XM_011511313.2
XP_011509615.1
3, 4
686
XM_011511315.2
XP_011509617.1
3, 4
686
XM_011511317.1
XP_011509619.1
3, 4
686
XM_011511318.1
XP_011509620.1
3, 4
686
XM_011511319.2
XP_011509621.1
3, 4
686
XM_011511322.1
XP_011509624.1
3, 4
686
XM_017004271.1
XP_016859760.1
3, 4
686
XM_017004272.1
XP_016859761.1
3, 4
686
XM_017004273.1
XP_016859762.1
3, 4
686
XM_017004274.1
XP_016859763.1
3, 4
686
XM_017004275.1
XP_016859764.1
3, 4
452
XM_017004276.1
XP_016859765.1
4, 5
773
XM_017004277.1
XP_016859766.1
3, 4
686
XM_017004278.1
XP_016859767.1
3, 4
686
XM_017004279.1
XP_016859768.1
3, 4
686
XM_017004280.1
XP_016859769.1
3, 4
686
XM_017004281.1
XP_016859770.1
2, 3
292
XM_017004282.1
XP_016859771.1
2, 3
536
XM_017004283.1
XP_016859772.1
4, 5
773
XM_017004284.1
XP_016859773.1
3, 4
686
XM_017004285.1
XP_016859774.1
3, 4
452
XM_017004286.1
XP_016859775.1
2, 3
536
XM_017004287.1
XP_016859776.1
2, 3
536
XM_017004288.1
XP_016859777.1
2, 3
536
XM_017004289.1
XP_016859778.1
2, 3
536
XM_017004290.1
XP_016859779.1
2, 3
536
XM_017004291.1
XP_016859780.1
4, 5
723
GenBank mRNA
AB034636.1
-
2, 3
269
AK000506.1
-
3, 4
469
AK225534.1
-
3, 4
469
AK296714.1
-
2, 3
255
AK314534.1
-
3, 4
486
AL136618.1
-
2, 3
248
BC033495.1
-
3, 4
502
BC073146.1
-
3, 4
443
5743806
CR936600.1
-
2, 3
237
CR936759.1
-
3, 4
410
KU178642.1
-
2, 3
344
KU178643.1
-
2, 3
344
Back To Top
More Information
Gene Ontology Categories:
Process(es)
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-f6n66:80/10.239.161.116:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0