Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL17RE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193380.1 | NP_001180309.1 | 4 | 416 | ||
NM_153480.1 | NP_705613.1 | 4 | 416 | |||
NM_153481.1 | NP_705614.1 | 2 | 280 | |||
NM_153483.2 | NP_705616.2 | 5 | 435 | |||
NR_104198.1 | 3 | 400 | ||||
XM_006712975.1 | XP_006713038.1 | 3 | 242 | |||
XM_006712976.1 | XP_006713039.1 | 3 | 380 | |||
XM_011533361.2 | XP_011531663.1 | 4 | 332 | |||
XM_011533362.2 | XP_011531664.1 | 4 | 351 | |||
XM_011533363.2 | XP_011531665.1 | 4 | 351 | |||
XM_011533364.2 | XP_011531666.1 | 4 | 351 | |||
GenBank mRNA | AF458069.1 | - | 4 | 416 | ||
AK074238.1 | - | 4 | 396 | |||
AK096793.1 | - | 3 | 378 | |||
AK096801.1 | - | 2 | 260 | |||
AK297697.1 | - | 4 | 382 | |||
AK314476.1 | - | 4 | 401 | |||
AY358451.1 | - | 4 | 416 | |||
BC035478.1 | - | 5 | 428 | |||
BC063110.1 | - | 4 | 414 | |||
BC137077.1 | - | 5 | 378 | 9021074 | ||
BC137078.1 | - | 5 | 378 | 9021075 |