Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPRH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161440.2 | NP_001154912.2 | 18 | 3250 | ||
NM_002842.4 | NP_002833.4 | 20 | 3784 | |||
XM_011527183.1 | XP_011525485.1 | 20 | 3805 | |||
XM_011527188.1 | XP_011525490.1 | 19 | 3754 | |||
XM_017027056.1 | XP_016882545.1 | 19 | 3792 | |||
XM_017027057.1 | XP_016882546.1 | 19 | 3780 | |||
XM_017027058.1 | XP_016882547.1 | 19 | 3825 | |||
XM_017027059.1 | XP_016882548.1 | 19 | 3744 | |||
XM_017027060.1 | XP_016882549.1 | 18 | 3744 | |||
XM_017027061.1 | XP_016882550.1 | 17 | 3258 | |||
XM_017027062.1 | XP_016882551.1 | 19 | 3700 | |||
XM_017027063.1 | XP_016882552.1 | 19 | 3693 | |||
GenBank mRNA | D15049.2 | - | 20 | 3758 |