Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ATXN2L SIRNA Assays ›
Gene Symbol:
ATXN2L
Gene Name:
ataxin 2 like
Gene Aliases:
A2D, A2LG, A2LP, A2RP
Chromosome Location:
Chr.16: 28823048 - 28837237 on Build GRCh38
UniGene ID:
Hs.460499
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
44646
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ATXN2L
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001308230.1
NP_001295159.1
4
634
NM_007245.3
NP_009176.2
4
634
NM_017492.3
NP_059867.3
4
634
NM_145714.2
NP_663760.1
4
634
NM_148414.2
NP_680780.1
4
634
NM_148415.2
NP_680781.1
4
634
NM_148416.2
NP_680782.1
4
634
XM_005255061.1
XP_005255118.1
4
634
XM_005255062.1
XP_005255119.1
4
634
XM_005255063.3
XP_005255120.1
4
634
XM_005255064.1
XP_005255121.1
4
634
XM_005255065.1
XP_005255122.1
4
634
XM_005255066.1
XP_005255123.1
4
634
XM_005255067.1
XP_005255124.1
4
634
XM_005255068.1
XP_005255125.1
4
634
XM_005255069.1
XP_005255126.1
4
634
XM_005255070.1
XP_005255127.1
4
634
XM_005255071.1
XP_005255128.1
4
634
XM_005255074.1
XP_005255131.1
4
634
XM_005255075.1
XP_005255132.1
4
634
XM_005255076.1
XP_005255133.1
4
634
XM_005255077.1
XP_005255134.1
4
517
XM_006721007.1
XP_006721070.1
4
634
XM_006721008.1
XP_006721071.1
4
634
XM_006721009.1
XP_006721072.1
4
634
XM_006721010.2
XP_006721073.1
4
634
XM_006721011.1
XP_006721074.1
4
634
XM_006721012.3
XP_006721075.1
4
634
XM_011545719.2
XP_011544021.1
4
634
XM_011545721.2
XP_011544023.1
4
634
XM_011545722.1
XP_011544024.1
4
334
XM_017022891.1
XP_016878380.1
4
517
XM_017022892.1
XP_016878381.1
4
334
GenBank mRNA
AF034373.1
-
6
736
AJ317970.1
-
4
590
AJ317971.2
-
4
590
AJ317972.2
-
4
590
AJ317973.2
-
4
590
AJ317974.2
-
4
590
AK289981.1
-
4
634
AY188334.1
-
6
736
AY188335.1
-
6
736
AY188336.1
-
6
736
AY188337.1
-
6
736
AY188338.1
-
6
736
BC068012.1
-
5
492
BC082760.1
-
4
430
5812230
BC136584.1
-
4
587
9020574
U70671.1
-
4
264
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cytoplasmic mRNA processing body assembly
stress granule assembly
cell-cell adhesion
protein binding
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-gk48b:80/10.239.167.222:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0