Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000800.4 | NP_000791.1 | 4 | 1572 | ||
NM_001144892.2 | NP_001138364.1 | 4 | 1092 | |||
NM_001144934.1 | NP_001138406.1 | 5 | 1285 | |||
NM_001144935.1 | NP_001138407.1 | 4 | 1191 | |||
NM_001257205.1 | NP_001244134.1 | 4 | 1482 | |||
NM_001257206.1 | NP_001244135.1 | 4 | 1479 | |||
NM_001257207.1 | NP_001244136.1 | 5 | 1225 | |||
NM_001257208.1 | NP_001244137.1 | 4 | 1223 | |||
NM_001257209.1 | NP_001244138.1 | 4 | 1238 | |||
NM_001257210.1 | NP_001244139.1 | 4 | 1222 | |||
NM_001257211.1 | NP_001244140.1 | 4 | 1220 | |||
NM_001257212.1 | NP_001244141.1 | 4 | 1089 | |||
NM_033136.3 | NP_149127.1 | 3 | 1468 | |||
NM_033137.2 | NP_149128.1 | 2 | 926 | |||
XM_005268390.4 | XP_005268447.1 | 4 | 1162 | |||
XM_011537607.2 | XP_011535909.1 | 5 | 1282 | |||
GenBank mRNA | X51943.1 | - | 3 | 1033 |