Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000764.2 | NP_000755.2 | 4 | 1139 | ||
NM_030589.2 | NP_085079.2 | 3 | 986 | |||
XM_017026383.1 | XP_016881872.1 | 4 | 4336 | |||
GenBank mRNA | BC117165.1 | - | 4 | 642 | 40125716 | |
BC143325.1 | - | 4 | 642 | 9051830 | ||
M33317.1 | - | 4 | 606 | |||
U22029.1 | - | 4 | 1140 |