Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2A13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000766.4 | NP_000757.2 | 7 | 1073 | ||
GenBank mRNA | AF209774.1 | - | 7 | 1052 | ||
AY513604.1 | - | 7 | 1052 | |||
AY513605.1 | - | 7 | 1052 | |||
AY513606.1 | - | 7 | 1052 | |||
AY513607.1 | - | 7 | 1052 | |||
AY513608.1 | - | 7 | 1052 | |||
AY513609.1 | - | 7 | 1052 |