Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002982.3 | NP_002973.1 | 2 | 207 | ||
GenBank mRNA | AK311960.1 | - | 2 | 199 | ||
AV733621.2 | - | 2 | 207 | |||
BC009716.1 | - | 2 | 190 | 3901489 | ||
BG530064.1 | - | 2 | 220 | |||
BT007329.1 | - | 2 | 134 | |||
M24545.1 | - | 2 | 203 | |||
M26683.1 | - | 1 | 72 | |||
M28226.1 | - | 2 | 180 | |||
S69738.1 | - | 2 | 165 | |||
S71513.1 | - | 2 | 187 | |||
X14768.1 | - | 2 | 187 |