Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PFKM SIRNA Assays ›
Gene Symbol:
PFKM
Gene Name:
phosphofructokinase, muscle
Gene Aliases:
ATP-PFK, GSD7, PFK-1, PFK1, PFKA, PFKX, PPP1R122
Chromosome Location:
Chr.12: 48105278 - 48146404 on Build GRCh38
UniGene ID:
Hs.75160
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
45
Purification
HPLC
Standard
In-Vivo Ready
Size
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
Availability
Inventoried
Catalog #
AM51331
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PFKM
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000289.5
NP_000280.1
4
362
NM_001166686.1
NP_001160158.1
6
664
NM_001166687.1
NP_001160159.1
4
216
NM_001166688.1
NP_001160160.1
3
305
XM_005268974.1
XP_005269031.1
7
561
XM_005268975.1
XP_005269032.1
7
700
XM_005268976.3
XP_005269033.1
7
743
XM_005268977.1
XP_005269034.1
6
547
XM_005268978.2
XP_005269035.1
6
460
XM_005268979.1
XP_005269036.1
6
547
XM_011538487.1
XP_011536789.1
6
495
XM_011538488.2
XP_011536790.1
4
327
XM_017019467.1
XP_016874956.1
7
630
XM_017019468.1
XP_016874957.1
6
482
XM_017019469.1
XP_016874958.1
6
547
GenBank mRNA
AK126020.1
-
9
765
AK126229.1
-
7
614
AK312284.1
-
4
244
BC000534.2
-
3
192
2964710
BC007798.2
-
3
172
BC012799.2
-
3
192
2964710
BC013298.2
-
3
192
2964710
BC021203.2
-
4
214
4100117
DA525437.1
-
4
362
DA560355.1
-
4
216
DA761651.1
-
3
305
DA795106.1
-
4
347
KU178251.1
-
3
177
KU178252.1
-
3
177
KU178253.1
-
3
177
M26066.1
-
4
253
U24183.1
-
4
253
Y00698.1
-
3
196
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_031715, D21869, BC094212, FQ211719, U25651, and others.
Panther Classification:
Molecular Function -
carbohydrate kinase
kinase
transferase
metabolite interconversion enzyme
Gene Ontology Categories:
Function(s)
Process(es)
glycogen catabolic process
fructose 6-phosphate metabolic process
glycolytic process
positive regulation of insulin secretion
glucose homeostasis
muscle cell cellular homeostasis
carbohydrate phosphorylation
protein oligomerization
canonical glycolysis
glycolysis from storage polysaccharide through glucose-1-phosphate
6-phosphofructokinase activity
protein binding
ATP binding
protein C-terminus binding
kinase binding
identical protein binding
protein homodimerization activity
metal ion binding
fructose binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-c4z4p:80/10.239.161.116:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0