Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GPS1 SIRNA Assays ›
Gene Symbol:
GPS1
Gene Name:
G protein pathway suppressor 1
Gene Aliases:
COPS1, CSN1, SGN1
Chromosome Location:
Chr.17: 82051477 - 82057470 on Build GRCh38
UniGene ID:
Hs.268530
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
46077
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GPS1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001321089.1
NP_001308018.1
3, 4
401
NM_001321090.1
NP_001308019.1
3, 4
401
NM_001321091.1
NP_001308020.1
3, 4
401
NM_001321092.1
NP_001308021.1
2, 3
197
NM_001321093.1
NP_001308022.1
3, 4
282
NM_004127.5
NP_004118.3
2, 3
197
NM_212492.2
NP_997657.1
2, 3
692
XM_005256350.1
XP_005256407.1
3, 4
369
XM_005256353.1
XP_005256410.1
3, 4
369
XM_005256359.1
XP_005256416.1
2, 3
161
XM_005256360.3
XP_005256417.1
2, 3
202
XM_011523569.1
XP_011521871.1
3, 4
222
XM_011523570.1
XP_011521872.1
3, 4
272
XM_017024524.1
XP_016880013.1
2, 3
179
XM_017024525.1
XP_016880014.1
3, 4
369
XM_017024526.1
XP_016880015.1
2, 3
179
XM_017024527.1
XP_016880016.1
2, 3
202
XM_017024528.1
XP_016880017.1
2, 3
145
XM_017024529.1
XP_016880018.1
2, 3
145
XM_017024530.1
XP_016880019.1
2, 3
145
XM_017024531.1
XP_016880020.1
3, 4
222
XM_017024532.1
XP_016880021.1
2, 3
161
XM_017024533.1
XP_016880022.1
2, 3
189
XM_017024534.1
XP_016880023.1
3, 4
222
XM_017024535.1
XP_016880024.1
3, 4
222
GenBank mRNA
AB209596.1
-
1, 2
801
AK026610.1
-
3, 4
244
AK222508.1
-
2, 3
162
AK225248.1
-
2, 3
159
AK289435.1
-
2, 3
315
AK297872.1
-
2, 3
173
BC000155.2
-
2, 3
153
2900944
BC064503.1
-
2, 3
604
6572943
BG775520.1
-
3, 4
260
BP306851.1
-
2, 3
197
BT009834.1
-
1, 2
82
U20285.2
-
2, 3
189
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
inactivation of MAPK activity
nucleotide-excision repair, DNA damage recognition
transcription-coupled nucleotide-excision repair
cell cycle
JNK cascade
COP9 signalosome assembly
cullin deneddylation
negative regulation of GTPase activity
GTPase inhibitor activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-kt7sl:80/10.239.164.98:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0