Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017631.5 | NP_060101.3 | 30, 31 | 4458 | ||
XM_011532103.2 | XP_011530405.1 | 30, 31 | 4458 | |||
GenBank mRNA | AK000042.1 | - | 2, 3 | 259 | ||
AK223184.1 | - | 2, 3 | 259 | |||
AK225191.1 | - | 2, 3 | 246 | |||
BC038115.2 | - | 30, 31 | 4403 | 5261001 | ||
BC074781.2 | - | 2, 3 | 237 | |||
BX648655.1 | - | 10, 11 | 1300 |