Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C9orf16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024112.3 | NP_077017.1 | 1, 2 | 232 | ||
GenBank mRNA | AK315753.1 | - | 1, 2 | 159 | ||
BC001857.2 | - | 1, 2 | 136 | 3529116 | ||
BC008887.2 | - | 1, 2 | 136 | 3027807 | ||
CN360246.1 | - | 1, 2 | 33 | |||
DB493059.1 | - | 1, 2 | 298 |