Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klrb1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173292.2 | NP_775414.2 | 2 | 169 | ||
GenBank mRNA | AF541943.1 | - | 2 | 169 | ||
EF100682.1 | - | 2 | 169 | |||
EF100684.1 | - | 2 | 169 | |||
FJ158034.1 | - | 2 | 169 | |||
U56936.1 | - | 355 |
Gene Symbol: Nkr-p1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040189.2 | NP_001035279.1 | 2 | 201 | ||
GenBank mRNA | AF525533.1 | - | 2 | 169 | ||
DQ157010.1 | - | 2 | 201 | |||
EF100678.1 | - | 2 | 169 | |||
JF739552.1 | - | 2 | 169 |