Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phex | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013004.1 | NP_037136.1 | 8 | 1371 | ||
GenBank mRNA | AJ001637.1 | - | 8 | 1371 |
Gene Symbol: LOC102554012 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762111.1 | XP_008760333.1 | 2 | 1248 |