Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppard | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013141.2 | NP_037273.2 | 6 | 767 | ||
XM_006256166.2 | XP_006256228.1 | 6 | 1579 | |||
XM_006256167.2 | XP_006256229.1 | 5 | 1686 | |||
XM_006256168.2 | XP_006256230.1 | 5 | 2581 | |||
XM_008772797.1 | XP_008771019.1 | 5 | 1814 | |||
GenBank mRNA | AJ306400.1 | - | 4 | 533 | ||
U40064.1 | - | 6 | 788 | |||
U75918.1 | - | 6 | 781 |