Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017096.3 | NP_058792.1 | 2 | 332 | ||
GenBank mRNA | AY310153.1 | - | 2226 | |||
AY318961.1 | - | 2226 | ||||
AY321318.1 | - | 2223 | ||||
AY321324.1 | - | 2226 | ||||
AY325196.1 | - | 2226 | ||||
AY325218.1 | - | 2226 | ||||
AY325232.1 | - | 2226 | ||||
BC091157.1 | - | 400 | 7368286 | |||
M83176.1 | - | 2 | 332 |