Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnj8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017099.4 | NP_058795.3 | 2, 3 | 558 | ||
GenBank mRNA | AB043636.1 | - | 1, 2 | 388 | ||
AB043637.1 | - | 1, 2 | 388 | |||
BC078863.1 | - | 541 | 7128284 | |||
D42145.1 | - | 2, 3 | 561 |