Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vmn2r51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105179.1 | NP_001098649.1 | 3 | 782 |
Set Membership: |
G-protein coupled receptor |