Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esp36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177587.1 | NP_001171058.1 | 4 | 1034 | ||
GenBank mRNA | AB307016.1 | - | 4 | 1033 |