Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gas2l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033331.2 | NP_001028503.2 | 11 | 6370 | ||
NM_001079876.1 | NP_001073345.1 | 11 | 6373 | |||
NM_001284344.1 | NP_001271273.1 | 12 | 5385 | |||
GenBank mRNA | AK018456.1 | - | 1 | 923 | ||
AK171905.1 | - | 2 | 3368 |