Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myrfl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033333.3 | NP_001028505.1 | 22 | 2611 | ||
GenBank mRNA | AK144383.1 | - | 22 | 2612 | ||
AK144411.1 | - | 22 | 2612 | |||
BC150915.1 | - | 22 | 2415 | 9087826 | ||
BC150923.1 | - | 22 | 2415 | 9087834 |