Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081027.2 | NP_001074496.2 | 28 | 3894 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY730969.1 | - | 548 | Chr. - - [-] on Build |