Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabbr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081141.1 | NP_001074610.1 | 7, 8 | 1521 |
Set Membership: |
G-protein coupled receptor |