Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029216.2 | NP_083492.2 | 23, 24 | 3927 | ||
GenBank mRNA | AK147563.1 | - | 23, 24 | 3928 |