Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Card9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037747.1 | NP_001032836.1 | 7 | 1141 | ||
GenBank mRNA | BC151023.1 | - | 7 | 1349 | 9087936 |