Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ano8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164679.1 | NP_001158151.1 | 13 | 2320 | ||
GenBank mRNA | AK173222.1 | - | 11 | 3308 | ||
BC027735.1 | - | 13 | 2222 | |||
BC027790.1 | - | 6 | 1794 | |||
BC042391.1 | - | 6 | 2515 | |||
BC053460.1 | - | 1 | 359 | |||
BC059855.1 | - | 4 | 853 |