Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prdm14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081209.2 | NP_001074678.2 | 8 | 1766 | ||
GenBank mRNA | AK134292.1 | - | 8 | 1790 | ||
AK141218.1 | - | 7 | 1544 | |||
KJ463838.1 | - | 7 | 1733 |