Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RGS6 SIRNA Assays ›
Gene Symbol:
RGS6
Gene Name:
regulator of G-protein signaling 6
Gene Aliases:
GAP
Chromosome Location:
Chr.14: 71932100 - 72632478 on Build GRCh38
UniGene ID:
Hs.509872
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
5059
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RGS6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001204416.2
NP_001191345.1
3, 4
477
NM_001204417.2
NP_001191346.1
3, 4
477
NM_001204418.2
NP_001191347.1
3, 4
477
NM_001204419.2
NP_001191348.1
3, 4
477
NM_001204420.2
NP_001191349.1
3, 4
477
NM_001204421.2
NP_001191350.1
3, 4
477
NM_001204422.2
NP_001191351.1
3, 4
477
NM_001204423.1
NP_001191352.1
2, 3
242
NM_001204424.1
NP_001191353.1
3, 4
715
NM_004296.6
NP_004287.3
3, 4
477
NR_135235.1
3, 4
477
XM_017021818.1
XP_016877307.1
3, 4
377
XM_017021819.1
XP_016877308.1
3, 4
377
XM_017021820.1
XP_016877309.1
2, 3
212
XM_017021821.1
XP_016877310.1
2, 3
212
XM_017021822.1
XP_016877311.1
2, 3
212
XM_017021823.1
XP_016877312.1
2, 3
212
XM_017021824.1
XP_016877313.1
3, 4
377
XM_017021825.1
XP_016877314.1
2, 3
212
XM_017021826.1
XP_016877315.1
2, 3
212
XM_017021827.1
XP_016877316.1
2, 3
212
XM_017021828.1
XP_016877317.1
2, 3
212
XM_017021829.1
XP_016877318.1
3, 4
377
XM_017021830.1
XP_016877319.1
2, 3
212
XM_017021831.1
XP_016877320.1
2, 3
212
XM_017021832.1
XP_016877321.1
2, 3
212
XM_017021833.1
XP_016877322.1
2, 3
212
GenBank mRNA
AB209462.1
-
3, 4
283
AF073920.2
-
3, 4
399
AF107619.1
-
2, 3
192
AF107620.1
-
2, 3
192
AF156932.1
-
2, 3
192
AK294486.1
-
3, 4
715
AK294895.1
-
2, 3
242
AK313481.1
-
3, 4
537
AY309097.1
-
2, 3
192
AY309098.1
-
2, 3
192
AY309099.1
-
2, 3
192
AY309100.1
-
2, 3
192
BC104877.1
-
3, 4
370
8143880
BC113572.1
-
3, 4
370
8322624
BC143699.1
-
3, 4
370
DA386861.1
-
1, 2
49
DC317315.1
-
477
DC343081.1
-
446
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_019342.
Gene Ontology Categories:
Function(s)
Process(es)
G-protein coupled receptor signaling pathway
regulation of G-protein coupled receptor protein signaling pathway
negative regulation of signal transduction
intracellular signal transduction
positive regulation of GTPase activity
signal transducer activity
GTPase activator activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-68wbn:80/10.239.160.160:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0