Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Isoc2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101598.1 | NP_001095068.1 | 5 | 640 | ||
GenBank mRNA | BC158139.1 | - | 5 | 540 | 9007402 |