Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bloc1s2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028607.1 | NP_082883.1 | 3 | 240 | ||
GenBank mRNA | AK010745.1 | - | 3 | 237 | ||
AK145313.1 | - | 3 | 241 | |||
BC048500.1 | - | 2 | 120 | |||
BC065806.1 | - | 3 | 233 | 1547643 |