Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019314.1 | NP_062187.1 | 3 | 942 | ||
XM_006254682.2 | XP_006254744.1 | 3 | 2495 | |||
XM_006254683.2 | XP_006254745.1 | 4 | 3568 | |||
GenBank mRNA | DQ137427.1 | - | 1 | 170 | ||
JN857942.1 | - | 3 | 942 | |||
U69882.1 | - | 3 | 942 |
Gene Symbol: LOC103690147 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772181.1 | XP_008770403.1 | 3 | 2497 | ||
XM_008772182.1 | XP_008770404.1 | 4 | 3526 |