Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C330007P06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029951.1 | NP_084227.1 | 7 | 2512 | ||
GenBank mRNA | AK032304.1 | - | 7 | 2425 | ||
AK035825.1 | - | 7 | 2501 | |||
AK036910.1 | - | 7 | 2433 | |||
AK079769.1 | - | 1 | 673 | |||
AK141615.1 | - | 7 | 2453 | |||
BC021479.1 | - | 2657 | 5344198 | |||
BC051144.1 | - | 7 | 2427 | 6588980 | ||
BC085302.1 | - | 7 | 2401 |