Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ywhab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019377.1 | NP_062250.1 | 2 | 271 | ||
GenBank mRNA | BC076502.1 | - | 2 | 265 | 7100393 | |
D17446.1 | - | 2 | 266 | |||
FQ214109.1 | - | 2 | 307 | |||
FQ229978.1 | - | 2 | 276 | |||
S55223.1 | - | 2 | 266 | |||
S83440.1 | - | 2 | 271 |