Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptma | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021740.1 | NP_068508.1 | 2 | 232 | ||
GenBank mRNA | BC061972.1 | - | 2 | 242 | 5623463 | |
BC092569.1 | - | 2 | 252 | 7307282 | ||
FQ210626.1 | - | 2 | 263 | |||
FQ212611.1 | - | 2 | 263 | |||
FQ214221.1 | - | 2 | 257 | |||
FQ215633.1 | - | 2 | 263 | |||
FQ219686.1 | - | 2 | 263 | |||
FQ220677.1 | - | 2 | 264 | |||
FQ221801.1 | - | 2 | 263 | |||
FQ225103.1 | - | 2 | 267 | |||
FQ226916.1 | - | 2 | 263 | |||
FQ226946.1 | - | 2 | 263 | |||
FQ227002.1 | - | 2 | 263 | |||
FQ227835.1 | - | 2 | 263 | |||
FQ229930.1 | - | 2 | 263 | |||
FQ231540.1 | - | 2 | 263 | |||
FQ231794.1 | - | 2 | 269 | |||
FQ232039.1 | - | 2 | 267 | |||
FQ233285.1 | - | 2 | 269 | |||
M20035.1 | - | 2 | 259 | |||
M33024.1 | - | 2 | 259 | |||
M60664.1 | - | 2 | 271 | |||
M60665.1 | - | 2 | 271 | |||
M86564.1 | - | 2 | 232 | |||
X55326.1 | - | 2 | 232 |
Gene Symbol: LOC100359583 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751278.3 | XP_003751326.1 | 1 | 269 |